Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ECMatrix™-221 E8 Laminin Substrate
ECMatrix™-221 E8 Laminin Substrate Xeno-free laminin-221 coating for the differentiation of pluripotent stem cells into cardiomyocytes
$696.13 - $1825.76
Specifications
Cell Type | Pluripotent stem cells into cardiomyocytes |
---|---|
Content And Storage | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
Substrate | Laminin substrate |
Storage Requirements | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
Substrate Quantity | 0.5 mg/mL in PBS |
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
CC1631050MI
|
MilliporeSigma™
CC1631050UG |
6 x 175 μg |
Each for $1,825.76
|
|
|||||
CC163350UMI
|
MilliporeSigma™
CC163350UG |
2 x 175 μg |
Each for $696.13
|
|
|||||
Description
- Cardiomyocytes are cardiac muscle cells making up the heart, responsible for generating contractile forces within the heart
- Laminin-221 binds to the integrin α7X2β1, located on the cell surface of pluripotent stem cells and is known to induce cardiac and skeletal muscle differentiation
Specifications
Pluripotent stem cells into cardiomyocytes | |
Laminin substrate | |
0.5 mg/mL in PBS |
2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light | |
2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
For research purposes only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title