Learn More
Description
- Human pluripotent stem cells (ES and iPS cells) express α6β1 as the major integrin species and can be maintained stably and expanded efficiently in feeder-free conditions on culture vessels coated with the binding partner laminin-511
- Animal-free, xeno-free format: Consistent from lot-to-lot with no prescreening required
- No plate precoating required: Save time by adding to media while passaging cells
- Supports single cell passaging without ROCKi: Great for CRISPR editing or clonal isolation
- Higher adhesion and growth rates: Get to your experiments faster
- Easy to handle: No chilling of cell culture consumables required
Specifications
Specifications
| Cell Type | Pluripotent stem cell cultures, CHO-S derived |
| Form | Liquid |
| Substrate | Laminin |
| Includes | 6 x 175 μg ECMatrix™-511 E8 Laminin Substrate (expressed in CHO-S cells) |
| Packaging | 6 x 175 μg |
| Storage Requirements | Store 2°C to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
| Substrate Quantity | 0.5 mg/mL in PBS |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.