Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ECMatrix™-511 Silk E8 Laminin Substrate
ECMatrix™-511 Silk E8 Laminin Substrate Xeno-free laminin-511 coating for feeder-free pluripotent stem cell cultures
$217.50 - $720.00
Specifications
Cell Type | Puripotent stem cell cultures |
---|---|
Content And Storage | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
Form | Liquid |
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Price | Quantity & Availability | |||||
CC161100MMI
|
MilliporeSigma™
CC161100ML |
100 mL |
Each for $217.50
|
|
|||||
CC1611050MI
|
MilliporeSigma™
CC1611050UG |
6 x 175 μg |
Each for $720.00
|
|
|||||
Description
- Human pluripotent stem cells (ES and iPS cells) express α6β1 as the major integrin species and therefore can be maintained stably and expanded efficiently in feeder-free conditions on culture vessels coated with the binding partner laminin-511
- Animal-free, xeno-free format: Consistent from lot-to-lot with no prescreening required
- No plate precoating required: Save time by adding to media while passaging cells
- Supports single cell passaging w/out ROCKi: Great for CRISPR editing or clonal isolation
- Higher adhesion and growth rates
- Easy to handle: No chilling of cell culture consumables required
Specifications
Puripotent stem cell cultures | |
2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light | |
Liquid |
For research purposes only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title