Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Eco Best Technology™ Disposable Nitrile Gloves
GREENER_CHOICE

Catalog No. 19020403 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-403 Large
19-020-400 X-Small
19-020-401 Small
19-020-402 Medium
19-020-404 X-Large
19-020-405 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-020-403 Supplier SHOWA™ Supplier No. 6112PFL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eco Best Technology™ (EBT) biodegradable nitrile disposable gloves provide comfort, dexterity and performance across a wide range of industrial applications.

  • Same performance as non-EBT nitrile disposable gloves
  • Eco Best Technology™ (EBT) achieved 82% biodegradation in 386 days
  • Decreased risk of allergies
  • Second-skin fit and feel
  • Recommended for industrial applications including food, pharmaceutical, utilities, construction, agricultural and automotive

Compliance

Data Testing is available upon request.

Specifications

Certifications/Compliance ASTM D5511-18, ASTM D5526
Certifications/Compliance (Europe) EN 374-2:2003, EN 374-1:2016/Type B, EN 374-5:2016
Applications Industrial
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Size Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6112PFL
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.