Learn More
Restek™ Eco-Cap GL-45 Bottle Tops
SDP
Catalog No. 06611815 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Thread Size (Metric):
78 mm
Catalog No. Thread Size (Metric)
06-611-815 78 mm
1 options
Catalog No. 06-611-815 Supplier Restek™ Supplier No. 27834
Economically protect mobile phases from evaporation and contamination, and labs from harmful vapors
  • Hub-Cap Adaptor (cat.# 26538) required for use of Eco-Caps on 4-liter bottles
  • Tubing is inserted, no threading for fittings on inside of the tubing ports
  • Includes Polypropylene Male Luer Plug
Specifications
For Use With (Application) GL-45 Threaded Bottles
Includes Polypropylene Male Luer Plug
Thread Size (Metric) 78 mm
Neck Finish 78 mm
Product Type Connector Cap (3-port)