Learn More
Bevco Precision Manufacturing Co Econo ESD Chair with Casters; Mid
Supplier:  Bevco Precision Manufacturing Co V830SHC
Encompass_Preferred
Bevco's Lexington ESD Series chairs deliver all of the outstanding ergonomic benefits at a lower price point. This reliable chair was built to meet the demands of daily use for environments that require some level of static discharge control but do not need to meet ANSI/ESD 12.1 - 2019 specifications. Static dissipative with a resistance of 105-109 ohms when measured from seat/back to grounding chain only.Fully adjustable contoured back has a bumper guard on the edges to protect the upholstery.A waterfall seat aids lower body circulation promoting overall wellness.The pneumatic height adjustment allows you to easily change your seat height from 21" - 28.5"Upholstered fabric meets CAL 117 standards.The chair features a sturdy 27" diameter black nylon base with a brass drag chain and adjustable chrome footring.Model comes standard with dual-wheel hard floor casters.
Catalog No. NC2398366
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.