Learn More
Eisco™ Economy Bulb Holder
SCI_ED
Catalog No. S27757 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27757 1 Ea.
1 options
Catalog No. S27757 Supplier Eisco™ Supplier No. PH1004
Add to Cart
Add to Cart
This durable, economic Bulb Holder accepts size E10 Screw Bulbs - Miniature Edison Screw (MES).
  • Single holder
  • Made of hard plastic with brass rivets
  • Securely holds socket and terminals to base
  • Accepts E10 Screw Bulbs - Miniature Edison Screw (MES)
  • Base Diameter: 2.16 in. (55 mm)
Specifications
Product Type Bulb Holder, Economy
Material Plastic
For Use With (Application) E10 Screw Bulbs - Miniature Edison Screw (MES)
Quantity 1 Ea.