Learn More
MilliporeSigma™ Supelco™ Economy Moisture Trap, Fitting Size 1/4 in.
Catalog No. 111016418 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
400 cc
Catalog No. Volume (Metric)
11-101-6418 400 cc
1 options
Catalog No. 11-101-6418 Supplier MilliporeSigma™ Supelco™ Supplier No. 23988
Add to Cart
Add to Cart
Designed for pneumatic control to drive equipment, such as autosamplers and valves.
The clear polycarbonate tubes contains a mixture of Molecular Sieve 13X and Molecular Sieve 4A with indicating capability. Because room air may permeate through the polycarbonate tube, this trap should not be used with carrier gas streams. It is designed for pneumatic control to drive equipment, such as autosamplers and valves. The indicating 4A changes from blue to tan at 20% relative humidity. Note: Not for use with carrier gas streams. Dimensions: . 400 cc Traps: 17 1/2 in. (44.4 cm) long x 2 in. (51 mm) diameter
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 400 cc