Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Economy Optics Kit
SDPSCI_ED

Catalog No. S65110 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S65110 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65110 Supplier United Scientific™ Supplier No. OPTKIT
Add to Cart
Add to Cart

An entire Optics Kit designed to generate 20 different activities and experiments.

  • Includes everything needed to demonstrate basic principles of light and color.
  • Activity guide explains 20 experiments.
  • For 1 to 2 lab groups.

Includes:

  • Double convex lens, 38 mm diameter, glass
  • Double concave lens, 38 mm diameter, glass
  • Plain lens, 38 mm diameter, glass
  • Clear glass screen
  • Two mirrors, acrylic, square
  • Three lens/mirror supports
  • Five cellophane squares in five colors
  • Two prisms, equilateral, 25 x 50 mm, acrylic
  • Beaker, graduated, plastic
  • Flashlight with batteries
  • Plastic screen, white
  • Candle
  • Storage box

Specifications

Grade 5+
For Use With (Application) Demonstrate laws of reflection and refraction in optics
Includes Activity Guide, Double Convex Lens, 38 mm dia., Glass, Double Concave Lens, 38 mm dia., Glass, Plain Lens, 38 mm dia., Glass, Clear Glass Screen, 2 Mirrors, Acrylic, Square, 3 Lens/mirror Supports, 5 Cellophane Squares In 5 Colors, 2 Prisms, Equilateral, 25
Product Type Economy Optics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.