Learn More
Eisco™ Economy Sling Psychrometer Kit
SCI_ED
Catalog No. S23758 Shop All Eisco Products
Change view
Click to view available options
Includes:
(1) Dry Bulb Thermometer, (1) Wet Bulb Thermometer, (1) Handle, (1) Moisture Wick Cloth, (1) Screw, (2) Rubber Bands, (1) Set of instructions with psychometric chart of relative humidity
Catalog No. Includes
S23758 (1) Dry Bulb Thermometer, (1) Wet Bulb Thermometer, (1) Handle, (1) Moisture Wick Cloth, (1) Screw, (2) Rubber Bands, (1) Set of instructions with psychometric chart of relative humidity
1 options
This item is not returnable. View return policy
Catalog No. S23758 Supplier Eisco™ Supplier No. PH1120142
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
(1) Dry Bulb Thermometer, (1) Wet Bulb Thermometer, (1) Handle, (1) Moisture Wick Cloth, (1) Screw, (2) Rubber Bands, (1) Set of instructions with psychometric chart of relative humidity
For determining relative atmospheric humidity.
  • Two thermometers attached to end of handle and moist cloth placed over one bulb
  • Swing apparatus to read difference in temperature between wet bulb and dry bulb
Specifications
Includes (1) Dry Bulb Thermometer, (1) Wet Bulb Thermometer, (1) Handle, (1) Moisture Wick Cloth, (1) Screw, (2) Rubber Bands, (1) Set of instructions with psychometric chart of relative humidity
For Use With (Application) determining relative atmospheric humidity
Product Type Psychrometer Kit