Learn More
Cole-Parmer™ Oakton™ EcoTestr™ pH 1 Waterproof Pocket Tester
SCI_ED
Catalog No. S81377 Shop All Oakton Products
Change view
Click to view available options
Range (pH):
0 to 14
Catalog No. Range (pH)
S81377 0 to 14
1 options
Catalog No. S81377 Supplier Cole-Parmer™ Supplier No. 3563405
Add to Cart
Add to Cart
Oakton™ EcoTestr™ pH 1 Waterproof Pocket Tester automatically recognizes pH buffer standards
  • Large LCD dual-line display shows pH and temperature readings simultaneously
  • Rugged IP67-rated housing is waterproof, dustproof, and floats in water
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure - attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
Specifications
Description Waterproof pH Tester
Accuracy (Temperature) ±0.5°C;/±0.9°F
Accuracy (pH) ±0.1
Includes Plastic Storage Case, Lanyard, AAA Batteries (4)
Measurement Modes pH
Calibration Automatic
Range (pH) 0 to 14
Resolution (pH) 0.1
Calibration Points Up to 3
Temperature Range (Metric) 0°C to 50°C
Display Type LCD, Large, Dual-Line
Backlight Option None
Material IP67-Rated Housing
Type pH Tester