Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium, Dihydrate, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18602392 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-392 45 kg Fiber Drum
18-602-385 125 g Poly Bottle
18-602-387 500 g Poly Bottle
18-602-386 6 x 125 g Poly Bottle
18-602-389 2.5 kg Poly Pail
18-602-388 6 x 500 g Poly Bottle
18-602-390 4 x 2.5 kg Poly Pail
18-602-391 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-602-392 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ED15045KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ED150, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Edetate Disodium, Dihydrate, USP is a chelating agent used to control heart rhythm disturbances caused by a heart medications. All Spectrum Chemical USP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 8.7-11.4%
Packaging Fiber Drum
Quantity 45 kg
Percent Purity 99 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.