Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium Dihydrogen, Dihydrate, Primary Standard, 99.9-100.1%, Spectrum™ Chemical
SDP

Catalog No. 18602377 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-377 25 g Poly Bottle
18-602-378 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-377 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E124725GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

E1247, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Edetate Disodium Dihydrogen, Dihydrate, Primary Standard is a single component organic chemical reference standard. Reference standards are particularly important for analytical chemistry since most analytical instrumentation is comparative; it requires a sample of known composition (reference standard) for accurate calibration. These reference standards are produced under stringent manufacturing procedures and differ from laboratory reagents in their certification and the traceability of the data provided.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Percent Purity 99.9 to 100.1%
Grade Primary Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.