Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium TS, 0.1% (w/v) in 5% (v/v) Alcohol, Ricca Chemical
SDP

Catalog No. 266416 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2664-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2664-16 Supplier Ricca Chemical Company Supplier No. 266416
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 6381-92-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
CAS Min % 94.0, 3.96, 0.1
CAS Max % 98.0, 4.12, 0.1
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix 5% (v/v) Alcohol
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.1% (w/v)
Chemical Name or Material Edetate Disodium, Test Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.