Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533493
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533493 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533493 Supplier Thermo Scientific Supplier No. PA533493
Only null left
Add to Cart
Add to Cart

EDG3 Polyclonal Antibody

EDG3 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene S1PR3
Gene Accession No. Q99500
Gene Alias AI132464; cardiac sphingosine-1-phosphate specific receptor; EDG3; EDG-3; Edg3l; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; FLJ37523; FLJ93220; G protein-coupled receptor, endothelial differentiation gene-3; Lpb3; Lysophospholipid receptor B3; MGC71696; S1P receptor 3; S1P receptor EDG3; S1P receptor Edg-3; S1P receptor S1P3; S1p3; S1PR3; sphingosine; Sphingosine 1-phosphate receptor 3; sphingosine 1-phosphate receptor Edg-3; sphingosine-1-phosphate receptor 3
Gene Symbols S1PR3
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human EDG3 / S1P3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.