Learn More
Invitrogen™ EDG6 Monoclonal Antibody (1), FITC

Description
EDG6 Monoclonal Antibody for Flow, Western Blot
Specifications
Specifications
| Antigen | EDG6 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 1 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.08% sodium azide |
| Gene | S1PR4 |
| Gene Accession No. | O95977 |
| Gene Alias | Edg6; EDG-6; Endothelial differentiation G-protein coupled receptor 6; endothelial differentiation G-protein-coupled receptor 6; endothelial differentiation, G protein coupled receptor 6; endothelial differentiation, G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 6; LPC1; Lysophospholipid receptor C1; S1P receptor 4; S1P receptor Edg-6; S1P(4) receptor; S1p4; S1PR4; SLP4; sphingosine 1-phosphate receptor 4; Sphingosine 1-phosphate receptor Edg-6; sphingosine-1-phosphate receptor 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.