Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ 48-703 Seamless Knit Polyethylene Gloves with Leather Palm Pad

Catalog No. p-7162810 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves combine cut, abrasion, and thermal resistance with a leather palm pad for additional cushion and support during metal stamping applications.

  • Seamless construction with a knit wrist provides a close fit for a high level of dexterity
  • Re-inforcement protects the user when handling hot or sharp objects
  • Enhanced grip capabilities in dry or slightly oily environments
  • Leather palm pad offers additional cushioning and protection
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Construction Seamless Knit
Coating Material Nitrile
Color Gray
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Polyethylene, Fiberglass
Product Type Seamless Knit Gloves
Size 7
Style Clute
EN388 Standard 4X43D
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, Metal Fabrication, and Oil/Gas Industries
Glove Cuff Style Knit
Palm Type Leather
Series EDGE™ 48-703
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.