Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ 48-919 Double Layer Nitrile Coated Oil Repellent Gloves

Catalog No. 19501060 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-060 6
19-501-061 7
19-501-064 10
19-501-065 11
19-501-066 12
19-501-062 8
19-501-063 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-501-060 Supplier Ansell Supplier No. 48919060
Add to Cart
Add to Cart
This item is not returnable. View return policy

Double layer nitrile coating keeps hands dry and comfortable when handling oily parts, while an elastic knit wrist ensures a snug fit.

  • Seamless polyester liner is comfortable and flexible
  • Double nitrile coating provides protection against industrial fluid exposure
  • Rough texture on the palm ensures a secure grip in oily environments
  • Ideal for workers who handle small and medium-sized components coated with oil or lubricant
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Red, Black
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Polyester
Product Type Dip-coated Gloves
Size 6
For Use With (Application) Automotive, Construction, Machinery/Equipment, and Metal Fabrication
Glove Cuff Color Gray
Glove Cuff Style Knit
Series 48-919
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.