Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ EDGE™ Apron

Catalog No. 19267946 Shop All Ansell Products
Change view
Click to view available options
28 in. x 36 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-267-946 28 in. x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-267-946 Supplier Ansell™ Supplier No. EW0521200
This item is not returnable. View return policy

Ansell™ EDGE™ Apron is for workers who rely on consistent industrial clothing for protection from workplace risks

  • Microporous laminate construction
  • Comfort and quality at an affordable price
  • Integrated tie back
  • Latex and silicone free
  • Applications:
    • Maintaining lab furniture and equipment
    • Packaging and labeling
    • Assembly of parts
    • Equipment maintenance and repair
    • Cleaning of plant and machines
    • Maintenance of plant and machines
    • Inspection
    • Final Assembly
    • Testing
TRUSTED_SUSTAINABILITY

Specifications

Type Apron
Material Microporous Laminate
Color White
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Construction Microporous laminate construction
Latex Free Yes
No. of Pockets None
Dimensions (L x W) 28 in. x 36 in.
Style EDGE 67-200 Series - Apron
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.