Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals BIGGY Nickerson Agar

Catalog No. EBP4065
Change view
Click to view available options
Color:
light amber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
EB-P4065 light amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4065 Supplier Edge Biologicals Supplier No. P4065
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for the presumptive identification of Candida species

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Color light amber
Media Type BIGGY Nickerson Agar
For Use With (Application) identification of Candida species
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.