Learn More
Edge Biologicals Dehydrated Culture Media: Baird Parker Agar
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Form | Dehydrated |
| Media Type | Baird-Parker Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Lithium Chloride 5g/L, Agar 20g/L, Deionized Water 950mL, Egg Yolk Tellurite Suspension 50mL |
| For Use With (Application) | Isolation and enumeration of coagulase positive staphylococci |
| Packaging Type | Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.