Learn More
Edge Biologicals Dehydrated Culture Media: BIGGY Agar
Specifications
Specifications
| Form | Dehydrated |
| Media Type | BIGGY Agar |
| Product Type | Agar |
| For Use With (Application) | Isolation of fungus |
| Packaging Type | Slant |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.