Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Blood Agar

Catalog No. EBT0096
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0096 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0096 Supplier Edge Biologicals Supplier No. T0096
Add to Cart
Add to Cart

For growth of wide variety of microorganisms and determining hemolytic reactions

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Blood Agar
Product Type Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L, Defibrinated Sheep Blood 50mL, Deionized Water 950mL
For Use With (Application) For growth of wide variety of microorganisms and determining hemolytic reactions
Packaging Type Slant
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.