Learn More
Edge Biologicals Dehydrated Culture Media: Brain Heart Infusion (BHI) Broth
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Form | Dehydrated |
| Media Type | Brain Heart Infusion (BHI) Broth |
| Product Type | Nutrient Broth |
| Formulation | Brain heart infusion 6g/L, Peptic digest of Animal Tissue 6g/L, Pancreatic digest of Gelatin 14.5g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Disodium Phosphate 2.5g/L, Deionized Water 1000mL |
| For Use With (Application) | Cultivation of a variety of bacteria, Yeast and fungi |
| Packaging Type | Tube |
| Quantity | 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.