Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Brain Heart Infusion (BHI) Broth

Catalog No. EBT0051
Change view
Click to view available options
Quantity:
1 mL
2 mL
4 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0051 4 mL
EB-T0046 2 mL
EB-T0050 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EB-T0051 Supplier Edge Biologicals Supplier No. T0051
Add to Cart
Add to Cart

For cultivation of variety of bacteria, Yeast and fungi

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Brain Heart Infusion (BHI) Broth
Product Type Nutrient Broth
Formulation Brain heart infusion 6g/L, Peptic digest of Animal Tissue 6g/L, Pancreatic digest of Gelatin 14.5g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Disodium Phosphate 2.5g/L, Deionized Water 1000mL
For Use With (Application) Cultivation of a variety of bacteria, Yeast and fungi
Packaging Type Tube
Quantity 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.