Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Brain Heart Infusion Broth w/Gentamicin

Catalog No. EBT0068
Change view
Click to view available options
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
EB-T0068 Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0068 Supplier Edge Biologicals Supplier No. T0068
Add to Cart
Add to Cart
This item is not returnable. View return policy

For fungus isolation

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Form Dehydrated
Media Type Brain Heart Infusion (BHI) Broth with Gentamicin
Product Type Microbiology Media
For Use With (Application) For fungus isolation
Includes Gentamicin
Packaging Type Tube
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.