Learn More
Edge Biologicals Dehydrated Culture Media: DNASE Agar
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Form | Dehydrated |
| Media Type | DNASE Agar |
| Product Type | Agar |
| For Use With (Application) | For bacterial differentiation |
| Packaging Type | Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.