Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Eosin Methylene Blue (EMB) Agar

Catalog No. EBP1180
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P1180 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P1180 Supplier Edge Biologicals Supplier No. 1P180
Add to Cart
Add to Cart
This item is not returnable. View return policy

For selective isolation and presumptive identification of gram negative bacilli

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Eosin Methylene Blue (EMB) Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L, Deionized Water 1000mL
For Use With (Application) For selective isolation and presumptive identification of gram negative bacilli
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.