Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Lactose Broth

Catalog No. EBT0291
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-T0291 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0291 Supplier Edge Biologicals Supplier No. T0291
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection of presence of Coliform organisms and determination of Lactose fermentation of bacteria

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8-25°C
Form Dehydrated
Media Type Lactose Broth
Product Type Microbiology Media
Formulation Beef extract 3g/L, Lactose 5g/L, Peptone 5g/L, Deionized Water 1000mL
For Use With (Application) Detection of presence of Coliform organisms and determination of Lactose fermentation of bacteria
Includes Durham tube
Packaging Type Durham Tube
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.