Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Middlebrook 7H11 Agar

Catalog No. ebt0361
Change view
Click to view available options
Packaging Type:
Slant Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
EB-T0361 Slant Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0361 Supplier Edge Biologicals Supplier No. T0361
This item is not returnable. View return policy

For mycobacteria detection

Specifications

Form Dehydrated
Media Type Middlebrook 7H10 Agar
Product Type Agar
For Use With (Application) For Mycobacteria detection
Packaging Type Slant Tube
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.