Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Prepared Culture Media: Motility Test Medium

Catalog No. EBT0396
Change view
Click to view available options
Motility Test Medium with TTC; 13 x 100mm; Container: Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0396 Motility Test Medium with TTC; 13 x 100mm; Container: Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0396 Supplier Edge Biologicals Supplier No. T0396

For bacterial differentiation

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Content And Storage 8°C to 12°C
Form Prepared Medium
Media Type Motility Test Medium
Product Type Motility Test Medium
Formulation Tryptose 10g/L, Sodium Chloride 5g/L, Agar 4g/L, Deionized Water 1000mL
For Use With (Application) For bacterial differentiation
Includes Triphenyltetrazolium Chloride
Packaging Type Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.