Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Mueller Hinton Agar

Catalog No. EBP4245
Change view
Click to view available options
For Use With (Application):
Test microbial susceptibility
Media Type:
Mueller Hinton Agar with Blood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Media Type
EB-P4245 Test microbial susceptibility Mueller Hinton Agar with Blood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4245 Supplier Edge Biologicals Supplier No. P4245

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For testing susceptibility of microorganisms to antimicrobial agents

Specifications

Form Dehydrated
Media Type Mueller Hinton Agar with Blood
Product Type Agar
For Use With (Application) Test microbial susceptibility
Includes Blood
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.