Learn More
Edge Biologicals Dehydrated Culture Media: Phenylalanine Agar
Catalog No. EBT0496
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
Catalog No. Certifications/Compliance
EB-T0496 Certificate of analysis
1 options
This item is not returnable. View return policy
Catalog No. EB-T0496 Supplier Edge Biologicals Supplier No. T0496
Add to Cart
Add to Cart
This item is not returnable. View return policy
For differentiation of members of Proteus, Morganella and Providencia groups
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Certifications/Compliance Certificate of analysis
Form Dehydrated
Media Type Phenylalanine Agar
Product Type Agar
For Use With (Application) For differentiation of the members of the Proteus, Morganella and Providencia groupsFor differentiation of the members of the Proteus, Morganella and Providencia groups
Packaging Type Slant