Learn More
Edge Biologicals Dehydrated Culture Media: Plate Count Agar
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Form | Dehydrated |
| Media Type | Plate Count Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L, Deionized Water 1000mL |
| For Use With (Application) | For enumeration of microbial plate counts |
| Packaging Type | Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.