Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Plate Count Agar

Catalog No. EBT0506
Change view
Click to view available options
Packaging Type:
Deep Tube
Plate
Slant
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
EB-T0506 Deep Tube
EB-P2285 Plate
EB-T0501 Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EB-T0506 Supplier Edge Biologicals Supplier No. T0506
Add to Cart
Add to Cart

For enumeration of microbial plate counts

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Plate Count Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L, Deionized Water 1000mL
Packaging Type Deep Tube
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.