Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Potato Dextrose Agar

Catalog No. EBP3290
Change view
Click to view available options
Packaging Type:
Deep Tube
Plate
Slant
For Use With (Application):
For fungus isolation
Yeast and molds cultivation
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type For Use With (Application)
EB-P3290 Plate Yeast and molds cultivation
EB-T0511 Slant For fungus isolation
EB-T0516 Deep Tube For fungus isolation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. EB-P3290 Supplier Edge Biologicals Supplier No. P3290
Add to Cart
Add to Cart
This item is not returnable. View return policy

For fungus isolation

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Potato Dextrose Agar
Product Type Agar
Formulation Potatoes 200g/L, Dextrose 20g/L, Agar 15g/L, Deionized Water 1000mL
For Use With (Application) Yeast and molds cultivation
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.