Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Sab Dex Agar

Catalog No. EBP4345
Change view
Click to view available options
Packaging Type:
Plate
Includes:
Gentamicin
Media Type:
Sab Dex Agar with Gentamicin
For Use With (Application):
Yeast and fungal isolation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Includes Media Type For Use With (Application)
EB-P4345 Plate Gentamicin Sab Dex Agar with Gentamicin Yeast and fungal isolation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P4345 Supplier Edge Biologicals Supplier No. P4345
Add to Cart
Add to Cart

For isolation of pathogenic Yeasts and fungi

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Description 15 x 100mm
Form Dehydrated
Media Type Sab Dex Agar with Gentamicin
Product Type Agar
For Use With (Application) Yeast and fungal isolation
Includes Gentamicin
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.