Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: SABHI Agar

Catalog No. EBP4305
Change view
Click to view available options
Packaging Type:
Plate
For Use With (Application):
Isolation of fungus
Description:
15 x 100mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type For Use With (Application) Description
EB-P4305 Plate Isolation of fungus 15 x 100mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4305 Supplier Edge Biologicals Supplier No. P4305
Add to Cart
Add to Cart
This item is not returnable. View return policy

For fungus isolation

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Description 15 x 100mm
Form Dehydrated
Media Type SABHI Agar
Product Type Agar
For Use With (Application) Isolation of fungus
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.