Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: SIM Medium

Catalog No. EBT0671
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0671 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0671 Supplier Edge Biologicals Supplier No. T0671
Add to Cart
Add to Cart

For determination of Hydrogen Sulfide production, indole formation and motility of enteric bacteria

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Form Dehydrated
Media Type SIM Medium
Product Type Motility Medium
For Use With (Application) Determination of Hydrogen Sulfide production, indole formation and motility of enteric bacteria
Packaging Type Tube
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.