Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Simmons Citrate

Catalog No. EBT0676
Change view
Click to view available options
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
EB-T0676 Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0676 Supplier Edge Biologicals Supplier No. T0676
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection of gram negative bacteria

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Form Dehydrated
Media Type Simmons Citrate
Product Type Media Supplement
For Use With (Application) Detection of gram negative bacteria
Packaging Type Slant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.