Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Thermal Agar

Catalog No. EBP4414
Change view
Click to view available options
Content And Storage:
8°C to 12°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
EB-P4414 8°C to 12°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4414 Supplier Edge Biologicals Supplier No. P4414
Add to Cart
Add to Cart
This item is not returnable. View return policy

To detect the production of thermal nucleases by staphylococci

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Thermal Agar
Product Type Agar
Formulation Agar 15g/L, Deoxyribonucleic Acid 300mg/L, Sodium Chloride 10g/L, TRIZ HCl 790mg/L, TRIZ base 5.47g/L, Calcium Chloride 1.1mg/L, Toluidine blue 91.74mg/L, Deionized Water 1000mL
For Use With (Application) To detect the production of thermal nucleases by staphylococci
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.