Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Tryptic Soy Broth

Catalog No. EBT0756
Change view
Click to view available options
Quantity:
4.5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0756 4.5 mL
EB-T0758 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EB-T0756 Supplier Edge Biologicals Supplier No. T0756
Add to Cart
Add to Cart

For cultivation of fastidious and non-fastidious microorganisms

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Tryptic Soy Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dextrose 2.5g/L, Dipotassium Phosphate 2.5g/L, Deionized Water 1000mL
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type Tube
Quantity 4.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.