Learn More
Edge Biologicals Dehydrated Culture Media: Tryptic Soy Broth
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Form | Dehydrated |
| Media Type | Tryptic Soy Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dextrose 2.5g/L, Dipotassium Phosphate 2.5g/L, Deionized Water 1000mL |
| For Use With (Application) | Cultivation of fastidious and non-fastidious microorganisms |
| Packaging Type | Tube |
| Quantity | 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.