Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: TT Broth

Catalog No. ebt0752
Change view
Click to view available options
:
TT Broth (Hajna); 16 x 125mm; 10mL; Container: Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0752 TT Broth (Hajna); 16 x 125mm; 10mL; Container: Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0752 Supplier Edge Biologicals Supplier No. T0752
This item is not returnable. View return policy

For selective enrichment method of isolating Salmonella typhi and other species of Salmonella from feces

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8-25°C
Form Dehydrated
Media Type TT Broth
Product Type Microbiology Media
Formulation Yeast extract 2g/L, Dextrose 0.5g/L, Sodium Desoxycholate 0.5g/L, Sodium Thiosulfate 38g/L, Brilliant green 0.01g/L, Tryptose 18g/L, Mannito 2.5g/L, Sodium Chloride 5g/L, Calcium Carbonate 25g/L, Deionized Water 1000mL
For Use With (Application) For selective enrichment method of isolating Salmonella typhi and other species of Salmonella from feces
Packaging Type Tube
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.