Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Urea Broth

Catalog No. EBT0775
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-T0775 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0775 Supplier Edge Biologicals Supplier No. T0775
Add to Cart
Add to Cart

For detection of Urease activity of bacteria, especially members of Proteus and Providencia species.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8-25°C
Form Dehydrated
Media Type Urea Broth
Product Type Microbiology Media
Formulation Yeast extract 0.1g/L, Monopotassium Phosphate 9.1g/L, Dipotassium Phosphate 9.5g/L, Urea 20g/L, Phenol red 0.01g/L, Deionized Water 1000mL
For Use With (Application) Detection of Urease activity of bacteria, especially members of Proteus and Providencia species
Packaging Type Tube (2 mL)
Quantity 10/Pk. (2 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.