Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Violet Red Bile Agar

Catalog No. EBP3415
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P3415 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P3415 Supplier Edge Biologicals Supplier No. P3415
Add to Cart
Add to Cart
This item is not returnable. View return policy

For enumerating Coliform organisms

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Violet Red Bile Agar
Product Type Agar
Formulation Peptone 7g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Crystal Violet 2mg/L, Yeast extract 3g/L, Lactose 10g/L, Neutral red 30mg/L, Agar 15g/L, Deionized Water 1000mL
For Use With (Application) For enumerating Coliform organisms
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.