Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Vogel Johnson Agar

Catalog No. EBP3420
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P3420 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P3420 Supplier Edge Biologicals Supplier No. P3420
Add to Cart
Add to Cart

For early detection of coagulase positive Staphylococcus aureus

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Form Dehydrated
Media Type Vogel Johnson Agar
Product Type Agar
For Use With (Application) For early detection of coagulase positive Staphylococcus aureus
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.