Learn More
Edge Biologicals Sodium Hippurate Broth
Catalog No. EBT0681
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
EB-T0681 5 mL
1 options
This item is not returnable. View return policy
Catalog No. EB-T0681 Supplier Edge Biologicals Supplier No. T0681
Add to Cart
Add to Cart
This item is not returnable. View return policy
For presumptive identification of hippurate hydrolyzing bacteria, group B Streptococci (Streptococcus agalactiae)
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Broth Type Sodium Hippurate
For Use With (Application) Presumptive identification of hippurate hydrolyzing bacteria, group B Streptococci (Streptococcus agalactiae) based on their ability to hydrolyze hippurate
No. per Pack 10
Quantity 5 mL