Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ Lab Coat

Catalog No. 19167948 Shop All Ansell Products
Change view
Click to view available options
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-167-948 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-948 Supplier Ansell Supplier No. EW20208S
This item is not returnable. View return policy

Ansell™ EDGE™ Lab Coats are for workers who rely on industrial clothing for protection from workplace risks

  • Microporous laminate construction
  • Snap front design with elasticated cuffs are low linting and anti-static
  • Strong and durable yet breathable comfort
  • 2 pockets
  • Latex and silicone free
  • Applications:
    • Maintaining lab furniture and equipment
    • Packaging and labeling
    • Assembly of parts
    • Equipment maintenance and repair
    • Cleaning of plant and machines
    • Inspection
    • Final assembly
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Microporous Film
Color White
Closure Type Snaps
Cuff Style Elastic
Style Knee-length
No. of Pockets 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.