Learn More
Cayman Chemical EdonrpIcmaleate 25mg
SDP
Supplier:  Cayman Chemical 3602625 MG
Encompass_Preferred
A flavonoid inhibits benzo[a]pyrene-induced AhR-dependent transcription in a reporter assay at 2 and 10 UM decreases LPS-induced production of TNF-a in THP-1 cells (IC50 47 7 Ug/ml) inhibits cell migration and invasion of HN-30 HNSCC cells at 10 UM
Catalog No. 50-291-1890
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.